Learn More
Sino Biological Recombinant Anti-S100A6 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50229R01620
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse S100A6 / Calcyclin (rM S100A6; Catalog#50229-MNAE; Met 1-Lys 89; NP_035443.1).
Catalog No. 50-255-3360