Learn More
Sino Biological Recombinant Mouse S100A8 Protein (His Tag)
Supplier:  Sino Biological 50228M08B100
Encompass_Preferred
A DNA sequence encoding the mouse S100A8 (NP_038678.1) (Met1-Glu89) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-3362