Learn More
BIOHIPPO Mouse S100B Microsample
SDP
Catalog No. 5000314778
Encompass
Mouse S100B Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of S100B in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 18 5 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse S100B Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse S100B Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse S100B biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14778 Supplier BIOHIPPO Supplier No. BHE15211895
Mouse S100B Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of S100B in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 18 5 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse S100B Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse S100B Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse S100B biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color