Learn More
Sino Biological Anti-SAMSN1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 107480T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Mouse SAMSN1, and purified by antigen affinity chromatography.
Catalog No. 50-240-8748