Learn More
Sino Biological Recombinant Anti-LIMPII/SR-B2 Antibody (APC), Rabbit Monoclonal
Supplier:  Sino Biological 50052R052A100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse LIMP-2 / SCARB2 / CD36L2 (rM LIMP-2 / SCARB2 / CD36L2; Catalog#50052-M08H; NP_031670.1; Met1-Thr432) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-255-3515