Learn More
Sino Biological Recombinant Anti-LIMPII/SR-B2 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50052R017100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse SCARB2 / LIMP2 (rM SCARB2; Catalog#50052-M08H; Met 1-Thr 432; NP_031670.1).
Catalog No. 50-255-3517