Learn More
Sino Biological Anti-LIMPII/SR-B2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50052RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse SCARB2 / LIMP2 (rM SCARB2; Catalog#50052-M08H; NP_031670.1; Met 1-Thr 432). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3509