Learn More
BIOHIPPO Mouse SCFR Microsample
SDP
Catalog No. 5000315854
Encompass
Mouse SCFR Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of SCFR in Serum plasma and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 062 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse SCFR Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse SCFR Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse SCFR biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15854 Supplier BIOHIPPO Supplier No. BHE15211998
Mouse SCFR Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of SCFR in Serum plasma and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 062 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse SCFR Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse SCFR Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse SCFR biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color