Learn More
Sino Biological Recombinant Anti-Uteroglobin Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50291R016100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Uteroglobin / SCGB1A1 (rM Uteroglobin / SCGB1A1; Catalog#50291-M08H; Q06318; Met1-Phe96).
Catalog No. 50-241-1447