Learn More
Sino Biological Anti-Uteroglobin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50291T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Uteroglobin / SCGB1A1 (Catalog#50291-M08H; Q06318; Met1-Phe96). Uteroglobin / SCGB1A1 specific IgG was purified by Mouse Uteroglobin / SCGB1A1 affinity chromatography.
Catalog No. 50-241-2008