Learn More
Biomatik Corporation Mouse Sclerostin (SOST) CLIA Kit
Supplier:  Biomatik Corporation EKU09972
Encompass_Preferred
Mouse Sclerostin (SOST) CLIA Kit for detecting SOST in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 13.7-10,000pg/ml. Sensitivity: 6.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q99P68. Gene ID: 74499. Target Synonyms: VBCH; Sclerosteosis. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8391