Learn More
Sino Biological Mouse SDC1 Protein 1MG (Sino Biological)
Supplier:  Sino Biological 50641M08H1
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse SDC1 (NP_035649.1) (Met 1-Glu 252) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-237-3420