Learn More
Sino Biological Anti-Syndecan-4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50726T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Syndecan-4 / SDC4 (Catalog#50726-M08H; O35988; Met1-Val146). Syndecan-4 / SDC4 specific IgG was purified by Mouse Syndecan-4 / SDC4 affinity chromatography.
Catalog No. 50-255-3112