Learn More
FUJIFILM BIOSCIENCES INC Mouse SDF-1 alpha (CXCL12), Recombinant Protein, Shenandoah Biotechnology, 1 mg vial
Supplier:  FUJIFILM BIOSCIENCES INC 200461MG
Encompass_Preferred
Stromal cell-derived factor-1 alpha (SDF-1 A), also called CXCL12a, is one of two SDF-1 splice variants made by a wide variety of cells upon stimulation by inflammatory cytokines such as TNF, IL-1, and LPS. SDF-1A signals through the G protein-coupled receptor CXCR4 to recruit activated leukocytes. Human and mouse SDF-1 A share 99% sequence identity.
Catalog No. 50-990-880