Learn More
American Research Products Inc Mouse Secretoglobin family 3A
Catalog No. 5000327982
Encompass
Mouse Secretoglobin family 3A member 2 (SCGB3A2) ELISA kit from CUSABIO Detection Range 9 4 pg/mL-600 pg/mL Sensitivity 2 35 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27982 Supplier American Research Products Inc Supplier No. CSBEL020819MO
Mouse Secretoglobin family 3A member 2 (SCGB3A2) ELISA kit from CUSABIO Detection Range 9 4 pg/mL-600 pg/mL Sensitivity 2 35 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich