Learn More
American Research Products Inc Mouse Secretoglobin family 3A
Supplier: American Research Products Inc CSBEL020818MO
Mouse Secretoglobin family 3A member 1 (SCGB3A1) ELISA kit from CUSABIO Detection Range 62 5 pg/ml -4000 pg/ml Sensitivity 15 6 pg/ml Sample serum plasma tissue homogenates Method Sandwich