Learn More
Biomatik Corporation Mouse Secretogranin II (SCG2) ELISA Kit
Supplier:  Biomatik Corporation EKU07230
Encompass_Preferred
Mouse Secretogranin II (SCG2) ELISA Kit for detecting SCG2 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q03517. Gene ID: 20254. Target Synonyms: SN; SgII; CHGC; Chromogranin C; Secretoneurin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5792