Learn More
Sino Biological Anti-E-Selectin/CD62E Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50736T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse E-Selectin / CD62e / SELE (Catalog#50736-M08H; NP_035475.1; Met1-Pro564). E-Selectin / CD62e / SELE specific IgG was purified by Mouse E-Selectin / CD62e / SELE affinity chromatography.
Catalog No. 50-240-9524