Learn More
Sino Biological Recombinant Anti-L-selectin/CD62L Antibody (PE), Rabbit Monoclonal
Supplier:  Sino Biological 50045R414P100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse SELL / CD62L / L-selectin (rM SELL / CD62L / L-selectin; Catalog#50045-M08H; NP_035476.1; Met 1-Asn 332) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-240-8165