Learn More
Sino Biological Recombinant Anti-L-selectin/CD62L Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50045R02320
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD62L / L-Selectin / SELL (rM CD62L / L-Selectin / SELL; Catalog#50045-M08H; NP_035476.1; Met1-Asn332).
Catalog No. 50-241-1326