Learn More
Sino Biological Anti-L-selectin/CD62L Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50045RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse SELL / CD62L extracellular domain (rM SELL; Catalog#50045-M08H; NP_035476.1; Met 1-Asn 332). SELL specific IgG was purified by mouse SELL affinity chromatography.
Catalog No. 50-241-1771