Learn More
Sino Biological Recombinant Mouse P-Selectin/CD62P Protein (His Tag), HPLC-verified 1
Supplier:  Sino Biological 50737M08H1
Encompass_Preferred
A DNA sequence encoding the mouse SELP (Q01102) (Met1-Ala709) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-224-7301