Learn More
Sino Biological Recombinant Anti-PSGL-1/CD162 Antibody (PE), Rabbit Monoclonal
Supplier:  Sino Biological 50770R001P100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse PSGL-1/CD162 (rM PSGL-1/CD162; Catalog#50770-MCCH; Q62170; Met1-Cys307) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-241-0650