Learn More
Sino Biological Mouse SELPLG MAb 25TS (Sino Biological)
Supplier:  Sino Biological 50770R001A25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse PSGL-1/CD162 (rM PSGL-1/CD162; Catalog#50770-MCCH; Q62170; Met1-Cys307) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-237-1814