Learn More
Sino Biological Recombinant Anti-Semaphorin 4D Antibody (APC), Rabbit Monoclonal
Supplier:  Sino Biological 50642R010A100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Semaphorin 4D / SEMA4D / CD100 (rM Semaphorin 4D / SEMA4D / CD100; Catalog#50642-M08H; NP_038688.2; Met1-Arg733) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-255-3178