Learn More
Biomatik Corporation Mouse Semaphorin 3E (SEMA3E) ELISA Kit
Supplier:  Biomatik Corporation EKU07263
Encompass_Preferred
Mouse Semaphorin 3E (SEMA3E) ELISA Kit for detecting SEMA3E in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P70275. Gene ID: 20349. Target Synonyms: SEMAH; M-SemaK; M-sema H; coll-5; Sema Domain,Immunoglobulin Domain (Ig) ,Transmembrane Domain (TM) and Short Cytoplasmic Domain 3E. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8768