Learn More
Biomatik Corporation Mouse Serine Palmitoyltransferase, Long Chain Base Subunit 1 (SPTLC1) ELISA Kit
Supplier:  Biomatik Corporation EKU07281
Encompass_Preferred
Mouse Serine Palmitoyltransferase, Long Chain Base Subunit 1 (SPTLC1) ELISA Kit for detecting SPTLC1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O35704. Gene ID: 268656. Target Synonyms: HSAN; HSAN1; HSN1; LBC1; LCB1; SPT1; SPTI; Hereditary Sensory Neuropathy,Type 1; Long chain base biosynthesis protein 1; Serine-palmitoyl-CoA transferase 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4913