Learn More
Biomatik Corporation Mouse Serine Peptidase Inhibitor Kazal Type 1 (SPINK1) ELISA Kit
Supplier:  Biomatik Corporation EKU12244
Encompass_Preferred
Mouse Serine Peptidase Inhibitor Kazal Type 1 (SPINK1) ELISA Kit for detecting SPINK1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P09036. Gene ID: 20730. Target Synonyms: PCTT; PSTI; TATI; Tumor-Associated Trypsin Inhibitor; Pancreatic secretory trypsin inhibitor; Tumor-associated trypsin inhibitor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7294