Learn More
American Research Products Inc Mouse Serine protease HTRA1 H
Catalog No. 5000327710
Encompass
Mouse Serine protease HTRA1 (HTRA1) ELISA kit from CUSABIO Detection Range 78 pg/mL-5000 pg/mL Sensitivity 19 5 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27710 Supplier American Research Products Inc Supplier No. CSBEL010901MO
Mouse Serine protease HTRA1 (HTRA1) ELISA kit from CUSABIO Detection Range 78 pg/mL-5000 pg/mL Sensitivity 19 5 pg/mL Sample serum plasma tissue homogenates Method Sandwich