Learn More
Biomatik Corporation Mouse Serine/Threonine Kinase 11 (STK11) ELISA Kit
Supplier:  Biomatik Corporation EKU07291
Encompass_Preferred
Mouse Serine/Threonine Kinase 11 (STK11) ELISA Kit for detecting STK11 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.051ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9WTK7. Gene ID: 20869. Target Synonyms: LKB1; PJS; Polarization-Related Protein LKB1; Peutz-Jeghers Syndrome; Liver kinase B1; Renal carcinoma antigen NY-REN-19. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6079