Learn More
American Research Products Inc Mouse Serotransferrin TF ELI
Catalog No. 5000327810
Encompass
Mouse Serotransferrin (TF) ELISA kit from CUSABIO Detection Range 15 6 ng/mL-1000 ng/mL Sensitivity 3 9 ng/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-27810 Supplier American Research Products Inc Supplier No. CSBEL023412MO
Mouse Serotransferrin (TF) ELISA kit from CUSABIO Detection Range 15 6 ng/mL-1000 ng/mL Sensitivity 3 9 ng/mL Sample serum plasma Method Sandwich