Learn More
Biomatik Corporation Mouse Serpin Peptidase Inhibitor,Clade B,Member 4 (SERPINB4) ELISA Kit
Supplier:  Biomatik Corporation EKU07451
Encompass_Preferred
Mouse Serpin Peptidase Inhibitor,Clade B,Member 4 (SERPINB4) ELISA Kit for detecting SERPINB4 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.060ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Z2G2. Target Synonyms: SCCA2; PI11; SPB4; Leupin; Serpin Peptidase Inhibitor,Clade B,Member 4; Peptidase Inhibitor 11. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8751