Learn More
Sino Biological Anti-Cortisol Binding Globulin/CBG Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50314RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse SerpinA6 (rM SerpinA6; Catalog#50314-M08H; Q06770; Met 1-Ala 397). SerpinA6 specific IgG was purified by Mouse SerpinA6 affinity chromatography.
Catalog No. 50-240-7821