Learn More
Sino Biological Mouse SerpinA6 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 50314M27HB100
Encompass_Preferred
A DNA sequence encoding the Mouse SerpinA6 (Q06770) (Met1-Ala397) was expressed with a C-terminal Avi tag followed by a His tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-237-1794