Learn More
Bioworld Mouse Serum (8-12 weeks of age), sterile, 20 mL
Supplier:  Bioworld 306110841
Encompass_Preferred
Mouse serum is collected from sexually mature mice. This material is certified to have been collected from healthy mice, under controlled conditions. All mice were inspected and found to be free of infectious disease.SpecificationsSterile-filteredShips on dry ice.
Catalog No. 50-254-194