Learn More
Medchemexpress LLC Mouse Serum Albumin | 50 MG
SDP
Catalog No. 5000425773
Encompass_Preferred
Mouse Serum Albumin, the most abundant protein in plasma, leaks into the brain when the blood-brain barrier is impaired. It induces an A1 astrocyte phenotype, promotes VLSFAs secretion, and causes neuronal lippoapoptosis. MSA-activated microglia trigger tau phosphorylation, contributing to decreased spatial learning and memory in mice. This product is useful for studying neurodegenerative diseases like Alzheimer's and frontotemporal dementia.
  • Reduces primary neuron branch length, end-point voxels, and branches.
  • Upregulates Elovl1 mRNA and protein levels in primary astrocytes.
  • Induces significant neuronal apoptosis.
  • Impairs spatial learning and memory in mice.
  • Promotes tau phosphorylation.
  • Applicable for studying neurodegenerative diseases.
Catalog No. 50-004-25773 Supplier Medchemexpress LLC Supplier No. HYNP020450MG
Mouse Serum Albumin, the most abundant protein in plasma, leaks into the brain when the blood-brain barrier is impaired. It induces an A1 astrocyte phenotype, promotes VLSFAs secretion, and causes neuronal lippoapoptosis. MSA-activated microglia trigger tau phosphorylation, contributing to decreased spatial learning and memory in mice. This product is useful for studying neurodegenerative diseases like Alzheimer's and frontotemporal dementia.
  • Reduces primary neuron branch length, end-point voxels, and branches.
  • Upregulates Elovl1 mRNA and protein levels in primary astrocytes.
  • Induces significant neuronal apoptosis.
  • Impairs spatial learning and memory in mice.
  • Promotes tau phosphorylation.
  • Applicable for studying neurodegenerative diseases.