Learn More
American Research Products Inc Mouse serum amyloid A 3 protei
Catalog No. 5000327136
Encompass
Mouse serum amyloid A-3 protein (SAA3) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-27136 Supplier American Research Products Inc Supplier No. CSBEL020658MO
Mouse serum amyloid A-3 protein (SAA3) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich