Learn More
American Research Products Inc Mouse serum amyloid A1 SAA1
Catalog No. 5000327378
Encompass
Mouse serum amyloid A1 (SAA1) ELISA kit from CUSABIO Detection Range 3 12 ng/ml - 200 ng/ml Sensitivity 0 78 ng/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27378 Supplier American Research Products Inc Supplier No. CSBEL020656MO
Mouse serum amyloid A1 (SAA1) ELISA kit from CUSABIO Detection Range 3 12 ng/ml - 200 ng/ml Sensitivity 0 78 ng/ml Sample serum plasma tissue homogenates Method Sandwich