Learn More
American Research Products Inc Mouse serum amyloid A2 SAA2
Catalog No. 5000327226
Encompass
Mouse serum amyloid A2 (SAA2) ELISA kit from CUSABIO Detection Range 7 8 ng/mL-500 ng/mL Sensitivity 1 95 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27226 Supplier American Research Products Inc Supplier No. CSBEL020657MO
Mouse serum amyloid A2 (SAA2) ELISA kit from CUSABIO Detection Range 7 8 ng/mL-500 ng/mL Sensitivity 1 95 ng/mL Sample serum plasma tissue homogenates Method Sandwich