Learn More
Rockland Immunochemicals Mouse Serum (sterile) 100 mL
SDP
Catalog No. 501943963
Encompass_Preferred
Mouse Serum is used as a supplement to cell culture media. Mouse Serum is also suitable for use as a component of bioassays immunoassays or enzyme assays. Mouse Serum provides a broad spectrum of macromolecules carrier proteins for lipoid substances and trace elements attachment and spreading factors low molecular weight nutrients and hormones and growth factors that promote cell growth and health. Be certain to maintain Good Cell Culture Practice and maintain sterility of cultures that require media supplementation. Mouse Serum is ideal for investigators in Cancer Immunology and Cell Biology research.
Catalog No. 50-194-3963 Supplier Rockland Immunochemicals Supplier No. D108000100
Mouse Serum is used as a supplement to cell culture media. Mouse Serum is also suitable for use as a component of bioassays immunoassays or enzyme assays. Mouse Serum provides a broad spectrum of macromolecules carrier proteins for lipoid substances and trace elements attachment and spreading factors low molecular weight nutrients and hormones and growth factors that promote cell growth and health. Be certain to maintain Good Cell Culture Practice and maintain sterility of cultures that require media supplementation. Mouse Serum is ideal for investigators in Cancer Immunology and Cell Biology research.