Learn More
Bioworld Mouse serum, sterile filtered, heat inactivated, 500 mL
Supplier:  Bioworld 306111463
Encompass_Preferred
Collected from 8-12 week-old sexually mature mice that are free of common parasites.Physiochemical constituents are measured to ensure lot-to-lot consistency.Sterility tested.Hemoglobin levels < 35 mg/dl.Sterile filtered to 0.2 µm.Storage Conditions: -20°C + 5°C.Usage:bioWORLD's products are supplied for LABORATORY RESEARCH USE ONLY.The product may not be used as a drug,agricultural or pesticidal product,food additive or as a household chemical.
Catalog No. 50-254-227