Learn More
American Research Products Inc Mouse seX hormone binding glob
Catalog No. 5000326688
Encompass
Mouse seX hormone-binding globulin SHBG ELISA Kit from CUSABIO Detection Range 1 3 ng/mL-50 ng/mL Sensitivity 0 33 ng/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-26688 Supplier American Research Products Inc Supplier No. CSBE08233M
Mouse seX hormone-binding globulin SHBG ELISA Kit from CUSABIO Detection Range 1 3 ng/mL-50 ng/mL Sensitivity 0 33 ng/mL Sample serum plasma Method Sandwich