Learn More
BIOHIPPO Mouse sFLT Microsample
SDP
Catalog No. 5000315070
Encompass
Mouse sFLT Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of sFLT in serum plasma tissue homogenates saliva cell lysates follicular fluid and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 28 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse sFLT Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse sFLT Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse sFLT biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15070 Supplier BIOHIPPO Supplier No. BHE15210536
Mouse sFLT Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of sFLT in serum plasma tissue homogenates saliva cell lysates follicular fluid and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 28 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse sFLT Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse sFLT Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse sFLT biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color