Learn More
Sino Biological Anti-14-3-3 sigma Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50576T62100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse 14-3-3 sigma/SFN (rM 14-3-3 sigma/SFN; Catalog#50576-MNCE; NP_061224.2; Met1-Ser248). 14-3-3 sigma/SFN specific IgG was purified by Mouse 14-3-3 sigma/SFN affinity chromatography.
Catalog No. 50-255-3211