Learn More
Sino Biological Mouse SFRP4 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 24447H18HNA100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse sFRP4 (rM sFRP4; Catalog#50053-M08H; NP_057896.1; Met 1-Ser 351). sFRP4 specific IgG was purified by mouse sFRP4 affinity chromatography.
Catalog No. 50-237-4456