Learn More
Sino Biological Recombinant Anti-SP-D/SFTPD Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50205R019100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse SFTPD (rM SFTPD; Catalog#50205-M08H; NP_033186.1; Met 1-Phe 374).
Catalog No. 50-240-8106