Learn More
Sino Biological Anti-SP-D/SFTPD Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50205RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse SFTPD (rM SFTPD; Catalog#50205-M08H; NP_033186.1; Met 1-Phe 374). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-9783