Learn More
Sino Biological Anti-SP-D/SFTPD Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50205T48100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse SP-D/SFTPD (rM SP-D/SFTPD; Catalog#50205-M08H; NP_033186.1; Met1-Phe374). SP-D/SFTPD specific IgG was purified by Mouse SP-D/SFTPD affinity chromatography.
Catalog No. 50-255-3380