Learn More
Raybiotech Inc Mouse Sonic Hedgehog N-Terminal (Shh-N) ELISA Kit
Supplier:  Raybiotech Inc ELMSHHNCL1
Encompass_Preferred
For cell and tissue lysate samples. Sensitivity: 5 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes mouse Shh-N.
Catalog No. 50-195-0133