Learn More
Biomatik Corporation Mouse Sideroflexin 1 (SFXN1) ELISA Kit
Supplier:  Biomatik Corporation EKU07331
Encompass_Preferred
Mouse Sideroflexin 1 (SFXN1) ELISA Kit for detecting SFXN1 in tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q99JR1. Gene ID: 14057. Target Synonyms: TCC; Tricarboxylate carrier protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7620